Learn More
Biolegend TotalSeq™-A0851 anti-mouse CD1d (CD1.1, Ly-38)
Supplier:  Biolegend 123529
Encompass_Preferred
TotalSeq™-A0851 anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-207-0541