Learn More
Biolegend TotalSeq™-B0406 anti-human CD304 (Neuropilin-1)
Supplier:  Biolegend 354529
Encompass_Preferred
TotalSeq™-B0406 anti-human CD304 (Neuropilin-1) [12C2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-7375