Learn More
Biolegend TotalSeq™-C0022 anti-human CD137L (4-1BB Ligand)
Supplier:  Biolegend 311519
Encompass_Preferred
TotalSeq™-C0022 anti-human CD137L (4-1BB Ligand) [5F4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-3528