Learn More
Cole-Parmer Digi-Sense Touchscreen 12 Channel Scanning Data Logging Benchtop Thermocouple Thermometer
Catalog No. 501961601 Shop All Cole-Parmer Products
Encompass_Preferred
This 12-channel scanning thermometer simultaneously monitors different temperature processes and provides real-time data on your PC with the supplied data acquisition software. The thermometer also functions as a data logger, storing readings in internal nonvolatile memory for later download to PC in CSV format. Visible alarm alert you when a channel logs a temperature exceeding the high or low set point. Thermocouple temperature probes are sold separately.
Catalog No. 50-196-1601 Supplier Cole-Parmer Supplier No. 9200002
This 12-channel scanning thermometer simultaneously monitors different temperature processes and provides real-time data on your PC with the supplied data acquisition software. The thermometer also functions as a data logger, storing readings in internal nonvolatile memory for later download to PC in CSV format. Visible alarm alert you when a channel logs a temperature exceeding the high or low set point. Thermocouple temperature probes are sold separately.