Learn More
Research Products International Corp Touch-Free Auto Open Waste Can, Stainless Steel, 12 Gallon, 14 x 10 Inches
SDP
Catalog No. 501977391
Encompass
Automatic open and close feature protects users from contact with waste container or its contents. Allows convenient waste disposal and is ideal for laboratories that use biohazard material. Auto sensing lid detects motion from a hand or solid object moves across the top of container. Lid opens, then closes automatically. Powered by four ''D'' cell batteries (not included) that provide up to 4,000 cycles. On-Off switch deactivates the automatic function. Designed for dependable and rugged use. Features durable ABS plastic lid.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7391 Supplier Research Products International Corp Supplier No. 560182
Automatic open and close feature protects users from contact with waste container or its contents. Allows convenient waste disposal and is ideal for laboratories that use biohazard material. Auto sensing lid detects motion from a hand or solid object moves across the top of container. Lid opens, then closes automatically. Powered by four ''D'' cell batteries (not included) that provide up to 4,000 cycles. On-Off switch deactivates the automatic function. Designed for dependable and rugged use. Features durable ABS plastic lid.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.