Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Touch Screen Dry Bath/Block Heater PROMO

Catalog No. 88870009PM Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Blocks:
2 (must be ordered separately)
4 (must be ordered separately)
Voltage:
100/120 V
Includes:
Dry bath, US style plug, block lifter
Heating Rate:
≤ 20 min. 30° to 130°C
≤ 25 min. 30° to 130°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Blocks Voltage Includes Heating Rate
88-870-009PM 4 (must be ordered separately) 100/120 V Dry bath, US style plug, block lifter ≤ 25 min. 30° to 130°C
88-870-008PM 2 (must be ordered separately) 100/120 V Dry bath, US style plug, block lifter ≤ 20 min. 30° to 130°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-870-009PM Supplier Thermo Scientific™ Supplier No. 88870009PM

missing translation for 'validMainframeMsg'

Achieve precise temperature stability and uniformity with our easy-to-use touch screen dry bath.

  • Advanced internal temperature sensing probe with outstanding temperature accuracy and control
  • Temperature deviation adjustment
  • Precise temperature control with PID circuit
  • Temperature self-calibration enables user to offset the temperature to desired value
  • Timer enables the user to accurately monitor the heating time
  • Password protection feature
  • Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting
  • Range of configuration to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications
  • Powder-coated steel body construction ensures durability
  • When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
  • Easy-to-use touch screen control displays time and temperature
  • 10 user setting programs, each supports up to 5 sequential steps of different temperatures and duration times
  • Built-in over temperature protection device for added safety

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation

Required accessory:

  • Unit must be ordered with block(s)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID digital
Holds 4 Blocks
Lighting LED
Material Powder-coated Steel
Plug Type US
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C (Ambient at 25°C)
Type Dry Bath
Fuse 250V 8A
Heating Rate ≤ 25 min. 30° to 130°C
Includes Dry bath, US style plug, block lifter
No. of Blocks 4 (must be ordered separately)
Temperature Accuracy ≤ ±0.5°C
Temperature Uniformity ≤ ±1°C
Timer 0 to 99 hr. 59 min. or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 535 x 200 x 100 mm (21.1 x 7.9 x 3.9 in.)
Length (English) 21.1 in.
Length (Metric) 535 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 11.24 lb.
Weight (Metric) 5.1 kg
Voltage 100/120 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.