Learn More
Stoelting™ Touch Test Sensory Probes - Replacement Evaluators
SDP
Catalog No. 10000270 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Replacement Evaluator 1.65
Replacement Evaluator 2.36
Replacement Evaluator 2.44
Replacement Evaluator 2.83
Replacement Evaluator 3.22
Replacement Evaluator 3.61
Replacement Evaluator 3.84
Replacement Evaluator 4.08
Replacement Evaluator 4.17
Replacement Evaluator 4.31
Replacement Evaluator 4.56
Replacement Evaluator 4.74
Replacement Evaluator 4.93
Replacement Evaluator 5.07
Replacement Evaluator 5.18
Replacement Evaluator 5.46
Replacement Evaluator 5.88
Replacement Evaluator 6.10
Replacement Evaluator 6.45
Replacement Evaluator 6.65
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-270 Replacement Evaluator 6.65
10-000-251 Replacement Evaluator 1.65
10-000-252 Replacement Evaluator 2.36
10-000-253 Replacement Evaluator 2.44
10-000-254 Replacement Evaluator 2.83
10-000-255 Replacement Evaluator 3.22
10-000-256 Replacement Evaluator 3.61
10-000-257 Replacement Evaluator 3.84
10-000-258 Replacement Evaluator 4.08
10-000-259 Replacement Evaluator 4.17
10-000-260 Replacement Evaluator 4.31
10-000-261 Replacement Evaluator 4.56
10-000-262 Replacement Evaluator 4.74
10-000-263 Replacement Evaluator 4.93
10-000-264 Replacement Evaluator 5.07
10-000-265 Replacement Evaluator 5.18
10-000-266 Replacement Evaluator 5.46
10-000-267 Replacement Evaluator 5.88
10-000-268 Replacement Evaluator 6.10
10-000-269 Replacement Evaluator 6.45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 10-000-270 Supplier Stoelting™ Supplier No. 58040
Add to Cart
Add to Cart
Semmes-Weinstein monofilaments provide an approximate logarithmic scale of actual force, and a linear scale of perceived intensity. They have a long history of effective use in clinical settings, and can be used to diagnose pathologies of hyper- or hypo-aesthesia. Subsets within the set of 20 probes distinguish pathologies on different parts of the body (foot, hand, lip, cheek,etc.).
  • Set of 20 monofilaments based on the Semmes-Weinstein monofilament set
  • Features retractable filaments to protect the filament
  • Allows the researcher to carry a few in a pocket
Operating Principle
When the tip of a fiber of given length and diameter is pressed against the skin at right angles, the force of application increases as long as the researcher continues to advance the probe, until the fiber bends. After the fiber bends, continued advance creates more bend, but not more force of application. This makes it possible for the researcher using a hand-held probe to apply a reproducible force, within a wide tolerance, to the skin surface.
Pre-Clinical Use
Rodents exhibit a paw withdrawal reflex when the paw is unexpectedly touched. The Touch Test™ Sensory Evaluator can be used on the Plantar surfaces of the foot of a rat or mouse. The animal indicates sensation by pulling back its paw. Individual filaments available.
Specifications
For Use With (Application) Neuroscience
Model Replacement Evaluator 6.65