Learn More
Moldex™ TouchFree EcoStation™ Starter Kit
SDP
Catalog No. 19384379 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Hearing Protection Style:
Meteors
Pura-Fit
Softies
SparkPlugs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hearing Protection Style
19-384-379 Meteors
19-384-376 Softies
19-384-377 SparkPlugs
19-384-378 Pura-Fit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-384-379 Supplier Moldex™ Supplier No. 6837
Add to Cart
Add to Cart
TouchFree EcoStation™ - A Hygienic Solution to Earplug Dispensing Hearing protection has never been easier to access. Simply place your hand under the automatic dispenser and a pair of earplugs will drop right into your palm.
  • Hygenic From Dispenser to Your Hand
    • Earplugs drop a pair at a time into workers' hands, so you don't have to worry about plugs falling on the floor or trays collecting dust and contaminants. When the EcoStation is empty simply dispose of it in the recycle bin and replace with an easy to install refill. Empty EcoStation containers are also 100% recyclable.
  • Increased Compliance and Cost Savings
    • Convenient, quick access and ease of use for more compliance. Plus, workers take only one pair at a time, so there is less waste. Less waste means more cost savings. Lower cost per pair than packaged earplugs.
  • Snap-In Refill
    • Easy to install refill, snaps in quickly, for fast changeover. The sealed refill helps prevent contamination.
Specifications
Size Universal
Hearing Protection Style Meteors
Holds 500 Pairs
Quantity 4/Cs.
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Dielectric No
Material Foam
Mounting Wall Mountable
Foldable Roll Down
Includes Mounting Hardware