Learn More
BEST SANITIZERS An All-In-One Hand Hygiene system, the BSI dispensers can be used for soaps and sanitizers. Compatible with liquids, foams, and gels. Using the universal 1.2L cartridge, the manual dispenser will hold the same bottle as its touchless counterpart.
Catalog No. NC3338780
Encompass_Preferred
An All-In-One Hand Hygiene system, the BSI dispensers can be used for soaps and sanitizers. Compatible with liquids, foams, and gels. Using the universal 1.2L cartridge, the manual dispenser will hold the same bottle as its touchless counterpart. 4/CS
Catalog No. NC3338780 Supplier BEST SANITIZERS Supplier No. BHC9000
An All-In-One Hand Hygiene system, the BSI dispensers can be used for soaps and sanitizers. Compatible with liquids, foams, and gels. Using the universal 1.2L cartridge, the manual dispenser will hold the same bottle as its touchless counterpart. 4/CS