Learn More
Lime Media Touchless Stainless Dispenser
Catalog No. 19043982
Change view
Click to view available options
Quantity:
16/Cs.
Catalog No. Quantity
19-043-982 16/Cs.
1 options
Catalog No. 19-043-982 Supplier Lime Media Supplier No. 197775SST
Choose a touchless hand sanitizing dispenser made of high-grade stainless steel, bulk refillable 40oz tank, non-drip pump mechanism, and a low battery indicator light.
  • Stainless steel construction
  • 40 oz. bulk-fill tank
  • Wall or stand mount applicable
  • All hardware included
  • For use of gel or liquid sanitizer
  • Sanitizing fluid viewing window
  • 4 AA batteries required (not included)
Specifications
Product Type Touch-free Sanitizer Dispenser
Material Stainless Steel
Quantity 16/Cs.
Sterility Non-sterile