Learn More
Lime Media Touchless White Dispenser
Catalog No. 19043984
Change view
Click to view available options
Quantity:
15/Cs.
Catalog No. Quantity
19-043-984 15/Cs.
1 options
Catalog No. 19-043-984 Supplier Lime Media Supplier No. 197775WPT
Choose a touchless hand sanitizing dispenser made of high-grade white ABS plastic, bulk refillable 34 oz. tank, non-drip pump mechanism, and a low battery indicator light.
  • White ABS plastic
  • 34 oz. bulk-fill tank
  • Wall or stand mount applicable
  • All hardware included
  • For use of gel or liquid sanitizer
  • Sanitizing fluid viewing window
  • 4 C-cell batteries required (not included)
Specifications
Product Type Touch-free Sanitizer Dispenser
Quantity 15/Cs.
Sterility Non-sterile