Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magid™ TouchMaster Lightweight Seamless Lisle Inspection Gloves
SDP

Catalog No. 19278490
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-278490 Men's
19-278489 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-278490 Supplier Magid™ Supplier No. 13651
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from a blend of cotton and polyester for a cool and comfortable fit. Over-edge hem ensures a secure fit, while also preventing the cuff from unraveling during laundering.

  • Machine-knit lisle glove fits like a dress glove
  • Seamless, reversible design for longer wear
  • Cotton/Polyester blend provides a cool and comfortable fit
  • Over-edge hem helps to prevent unraveling when laundered
  • Ideal for inspection, or any general purpose applications
  • Color-coded hems for easy size identification

Specifications

Construction Machine Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polycotton
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
Style Reversible
For Use With (Application) Inspection, General Purpose Use
Gender Men's
Hem Color White
Series TouchMaster 1365
Length (English) 7.5 in.
Length (Metric) 19 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.