Learn More
Formlabs Inc TOUGH 2000 RESIN 1 L
SDP
Catalog No. NC1824639
Encompass
Whether you’re optimizing your manufacturing process, rapidly iterating through designs, or assessing form, fit, and function, Formlabs Engineering Resins are formulated to withstand extensive testing and perform under stress.Tough 2000 Resin is the strongest and stiffest material in our functional family of Tough and Durable Resins. Choose Tough 2000 Resin for prototyping strong and sturdy parts that should not bend easily.•Strong and stiff prototypes•Jigs and fixtures requiring minimal deflection •Simulating the strength and stiffness of ABSThe “2000” in the name represents the material’s tensile modulus. If you need a less stiff material, try Tough 1500 Resin or Durable Resin.
Catalog No. NC1824639 Supplier Formlabs Inc Supplier No. RSF2TO2001
Whether you’re optimizing your manufacturing process, rapidly iterating through designs, or assessing form, fit, and function, Formlabs Engineering Resins are formulated to withstand extensive testing and perform under stress.Tough 2000 Resin is the strongest and stiffest material in our functional family of Tough and Durable Resins. Choose Tough 2000 Resin for prototyping strong and sturdy parts that should not bend easily.•Strong and stiff prototypes•Jigs and fixtures requiring minimal deflection •Simulating the strength and stiffness of ABSThe “2000” in the name represents the material’s tensile modulus. If you need a less stiff material, try Tough 1500 Resin or Durable Resin.