Learn More
Fine Science Tools Fine Scissors, Tungsten Carbide & ToughCut, 11.5 cm
SDP
Catalog No. NC9839762
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: Tungsten Carbide & ToughCut
Catalog No. NC9839762 Supplier Fine Science Tools Supplier No. 1455911
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: Tungsten Carbide & ToughCut