Learn More
Fine Science Tools TOUGH CUT METZENBAUM SCISSORS,
SDP
Catalog No. NC0408797
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 29mm/ Alloy / Material: Stainless Steel/ Length: 18cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC0408797 Supplier Fine Science Tools Supplier No. 1411718
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 29mm/ Alloy / Material: Stainless Steel/ Length: 18cm/ Serrated: Yes/ Feature: ToughCut