Learn More
Fine Science Tools Surgical Scissors, ToughCut, Stainless Steel, 17.5 cm
SDP
Catalog No. NC9531910
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 60mm/ Alloy / Material: Stainless Steel/ Length: 17.5cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9531910 Supplier Fine Science Tools Supplier No. 1413017
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 60mm/ Alloy / Material: Stainless Steel/ Length: 17.5cm/ Serrated: Yes/ Feature: ToughCut