Learn More
Fine Science Tools Metzenbaum Scissors - ToughCut 
SDP
Catalog No. NC9788449
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 35mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9788449 Supplier Fine Science Tools Supplier No. 1411714
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 35mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: Yes/ Feature: ToughCut