Learn More
Electron Microscopy Sciences Tough-Spots Labels on Rolls, 1/2", Red
SDP
Catalog No. 5032719
Encompass_Preferred
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ½"
Color: Red
Catalog No. 50-327-19 Supplier Electron Microscopy Sciences Supplier No. 77581R
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ½"
Color: Red