Learn More
Andwin Scientific TOUGH SPOTS 1/2 YELLOW 1000/P
SDP
Catalog No. NC9482865
Encompass_Preferred
Tough-Spots Roll 1.5 u 2.0mL Tubes Yellow 1/2 in. (13mm) 1000/Roll - chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals. These labels withst
Catalog No. NC9482865 Supplier Andwin Scientific Supplier No. L60112
Tough-Spots Roll 1.5 u 2.0mL Tubes Yellow 1/2 in. (13mm) 1000/Roll - chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals. These labels withst