Learn More
Andwin Scientific TOUGH SPOTS 1/2IN BLUE 1000PK
SDP
Catalog No. NC9482866
Encompass_Preferred
Tough-Spots Roll 1.5 u 2.0mL Tubes Blue 1/2 in. (13mm) 1000/Roll - chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals. These labels withstan
Catalog No. NC9482866 Supplier Andwin Scientific Supplier No. L60116
Tough-Spots Roll 1.5 u 2.0mL Tubes Blue 1/2 in. (13mm) 1000/Roll - chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals. These labels withstan