Learn More
Andwin Scientific TOUGH SPOTS 1/2IN WHITE
SDP
Catalog No. NC9879429
Encompass_Preferred
The white Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2".
Catalog No. NC9879429 Supplier Andwin Scientific Supplier No. TSPOTS50
The white Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2".