Learn More
Andwin Scientific 3/8" Tough-Spots 1,000/roll, Lavender
SDP
Catalog No. NC1663647
Encompass_Preferred
Circular cryogenic labels formatted for handwriting. Tough Spots on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers. Tough Spots come in a variety of colors and sizes to ensure the right label for the right job!
Catalog No. NC1663647 Supplier Andwin Scientific Supplier No. TSPOTSL
Circular cryogenic labels formatted for handwriting. Tough Spots on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers. Tough Spots come in a variety of colors and sizes to ensure the right label for the right job!