Learn More
Electron Microscopy Sciences Tough-Spots Labels on Rolls, 3/8", Maroon
SDP
Catalog No. 5032712
Encompass_Preferred
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Maroon
Catalog No. 50-327-12 Supplier Electron Microscopy Sciences Supplier No. 77580MA
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Maroon