Learn More
Electron Microscopy Sciences Tough-Spots Tough-Spots on Roll, Pink, 3/8", 1000/roll
SDP
Catalog No. 501929692
Encompass_Preferred
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Pink
Catalog No. 50-192-9692 Supplier Electron Microscopy Sciences Supplier No. 77580P
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Pink