Learn More
Andwin Scientific TOUGH SPOTS 3/8IN YEL M/RL
SDP
Catalog No. NC9815939
Encompass_Preferred
The yellow Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196�C to 80�C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".
Catalog No. NC9815939 Supplier Andwin Scientific Supplier No. TSPOTSY
The yellow Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196�C to 80�C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".