Learn More
Electron Microscopy Sciences Tough-Spots Labels on Rolls, 7/16", White
SDP
Catalog No. 5032724
Encompass_Preferred
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: 7/16"
Color: White
Catalog No. 50-327-24 Supplier Electron Microscopy Sciences Supplier No. 7758044
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: 7/16"
Color: White