Learn More
Andwin Scientific TOUGH SPOTS BLUE 3/8IN
SDP
Catalog No. NC9333901
Encompass
The 3/8 in. Tough-Spots can withstand boiling, freezing, organic solvents and caustic agents. These are chemically inert labels that adhere to most plastic, glass and metals. It is available in color blue.
Catalog No. NC9333901 Supplier Andwin Scientific Supplier No. L10006B
Add to Cart
Add to Cart
The 3/8 in. Tough-Spots can withstand boiling, freezing, organic solvents and caustic agents. These are chemically inert labels that adhere to most plastic, glass and metals. It is available in color blue.