Learn More
USA Scientific Inc Tough-Spots, 3/8 inch diameter, coffee, on rolls
Catalog No. NC1315737
Encompass
Tough-Spots, small, coffee labels for 0.5 mL tubes, 3/8inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC1315737 Supplier USA Scientific Inc Supplier No. 91850017
Tough-Spots, small, coffee labels for 0.5 mL tubes, 3/8inch diameter, 1000/roll. For -196C to +80C.