Learn More
Andwin Scientific Tough-Spots, 3/8'', Green, 1000/roll
SDP
Catalog No. NC1663621
Encompass_Preferred
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.
Catalog No. NC1663621 Supplier Andwin Scientific Supplier No. TSPOTSG
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.