Learn More
USA Scientific Inc Tough-Spots, 3/8 inch diameter, green, on rolls
Catalog No. NC9583176
Encompass
Tough-Spots, small, green labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC9583176 Supplier USA Scientific Inc Supplier No. 91850002
Tough-Spots, small, green labels for 0.5 mL tubes, 3/8 inch diameter, 1000/roll. For -196C to +80C.