Learn More
Andwin Scientific 1/2" Tough-Spots, Silver, 1,000/roll
SDP
Catalog No. NC1663626
Encompass_Preferred
Tough-Spots are chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals.
Catalog No. NC1663626 Supplier Andwin Scientific Supplier No. TSPOTS50S
Tough-Spots are chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals.