Learn More
USA Scientific Inc Tough-Spots, 1/2 inch diameter, green, on rolls
Catalog No. NC9503221
Encompass
Tough-Spots, large, green labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC9503221 Supplier USA Scientific Inc Supplier No. 91850502
Tough-Spots, large, green labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.