Learn More
USA Scientific Inc Tough-Spots, 1/2 inch diameter, pink, on rolls
Catalog No. NC9228365
Encompass
Tough-Spots, large, pink labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC9228365 Supplier USA Scientific Inc Supplier No. 91850507
Tough-Spots, large, pink labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.