Learn More
Andwin Scientific TOUGH SPOTS WHITE 1/2IN
SDP
Catalog No. NC9875217
Encompass_Preferred
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents
Catalog No. NC9875217 Supplier Andwin Scientific Supplier No. L10008
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents