Learn More
Andwin Scientific TOUGH SPOTS WHITE 3/8IN 1000PK
SDP
Catalog No. NC9902145
Encompass_Preferred
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents
This item is not returnable. View return policy
Catalog No. NC9902145 Supplier Andwin Scientific Supplier No. L60102
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents