Learn More
Andwin Scientific Tough-Spots, 7/16'', White, 1000/roll
SDP
Catalog No. NC1663633
Encompass_Preferred
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.
Catalog No. NC1663633 Supplier Andwin Scientific Supplier No. TSPOTS44
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.