Learn More
Andwin Scientific TOUGH SPOTS YELLOW 3/8IN
SDP
Catalog No. NC9333899
Encompass
The 3/8 in. Tough-Spots can withstand boiling, freezing, organic solvents and caustic agents. These are chemically inert labels that adhere to most plastic, glass and metals. It is available in color yellow.
Catalog No. NC9333899 Supplier Andwin Scientific Supplier No. L10006Y
The 3/8 in. Tough-Spots can withstand boiling, freezing, organic solvents and caustic agents. These are chemically inert labels that adhere to most plastic, glass and metals. It is available in color yellow.