Learn More
Andwin Scientific TOUGH TAG YELLOW 1000/ROLLTO
SDP
Supplier:  Andwin Scientific 62045Y
Encompass_Preferred
The large yellow Tough-Tags labels are conveniently pre-cut to be easily put on microcentrifuge tubes and other laboratory containers. The labels can resist temperatures as low as -20C (-80C when used on virgin polypropylene tubes.) This product can also withstand organic solvents and caustic agents without peeling or falling off. The labels measure 1.28 in W x 0.5 in H, which are ideal for use with 1.5 mL tubes.
Catalog No. 50-996-480