Learn More
Electron Microscopy Sciences Tough-Tag Thermal Transfer Labels, 1.28 x 0.50", 3" Core
SDP
Supplier:  Electron Microscopy Sciences 7759920
Encompass_Preferred
Pre-cut, peel-off labels are designed for use in thermal transfer printers. A reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles, and other laboratory containers. Resists Degradation from scuffing, chemicals, moisture, and wide temperature fluctuations. Print as little as one label at a time, eliminating label waste and saving time and money. Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance. Specially coated for use with a resin ribbon that provides the best performance under extreme conditions. Print crisp, clear, professional quality labels.
Size: 1.28 x 0.50"
Core: 3"
Catalog No. 50-190-2273