Learn More
Labsource Inc TOUGH TAGS 1.28INX.5IN 1000/PK
SDP
Supplier:  Labsource Inc L60006PK
Encompass
Tough-Tags,1000 / Roll,Boil-/Freeze-proof ,Scuff and smear resistant ,Sized for 0.5ml or 1.5ml tubes ,Centrifuge compatable Tough-Tags are the pre-cut,peel-off,Label Size :1.28 x 0.5 in. ,Fits :1.5ml tubes
Catalog No. NC9896358