Learn More
Research Products International Corp Tough-Tags, Sidewall, Laser, 1/4x1 1/2"", Yellow, 3120/pk
SDP
Supplier:  Research Products International Corp 247137Y
Encompass
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to cryogenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies and Cryo-Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150°C.
Cryogenic labels formatted for laser printers.
Catalog No. 50-197-8059