Learn More
Electron Microscopy Sciences Tough-Tag Labels 22 x 22 mm, Rainbow
SDP
Supplier:  Electron Microscopy Sciences 775236
Encompass_Preferred
Labels for microcentrifuge tubes and other containers are temperature resistant from -40 degree C to 121 degree C and can be used in boiling water baths, autoclaving, and moderate freezer temperatures. Chemically inert, polyester sheets resist most organic solvents and caustic agents. They adhere to most plastics, glass and metals without cracking, peeling or degrading. Tubes labeled with Tough-Tags easily slide in and out of centrifuge rotors without binding. Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers.
Size: 22 x 22 mm
Color: Rainbow
Use: General Purpose
Catalog No. 50-326-65