Learn More
USA Scientific Inc Tough-Tags, 1.28 inch x 0.5 inch, yellow, on rolls
Supplier:  USA Scientific Inc 91871006
Encompass
Tough-Tags, large, yellow labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -40C to +121C.
Catalog No. NC9404752