Learn More
Fine Science Tools Fine Scissors - ToughCut, 9cm
SDP
Catalog No. NC9188278
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 20mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9188278 Supplier Fine Science Tools Supplier No. 1405909
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 20mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: ToughCut