Learn More
Fine Science Tools Fine Scissors, ToughCut, Stainless Steel, 11.5 cm
SDP
Catalog No. NC9955116
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9955116 Supplier Fine Science Tools Supplier No. 1405911
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes/ Feature: ToughCut