Learn More
Fine Science Tools Metzenbaum Scissors, ToughCut, 14.5 cm
SDP
Catalog No. NC9931142
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 35mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9931142 Supplier Fine Science Tools Supplier No. 1411614
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 35mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: Yes/ Feature: ToughCut