Learn More
Fine Science Tools TOUGHCUT VANNAS SPRING SCISSOR
SDP
Catalog No. NC0989941
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.3mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC0989941 Supplier Fine Science Tools Supplier No. 1512412
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.3mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm/ Serrated: Yes/ Feature: ToughCut