Learn More
STRATASYS INC TOUGHONE UPGRADE KIT FOR J850 SYSTEMS - Left UV Fan Duct.
Catalog No. NC3887871
Encompass
TOUGHONE UPGRADE KIT FOR J850 SYSTEMS - Left UV Fan Duct.Using ToughONE™ requires installing the new left air duct to ensure improved airflow throughout the printer. This upgrade is required for the J850 systems, including J850Pro, J850 Prme, J835, J850DAP, and J850FABRIX.
Catalog No. NC3887871 Supplier STRATASYS INC Supplier No. OBJ18097V
TOUGHONE UPGRADE KIT FOR J850 SYSTEMS - Left UV Fan Duct.Using ToughONE™ requires installing the new left air duct to ensure improved airflow throughout the printer. This upgrade is required for the J850 systems, including J850Pro, J850 Prme, J835, J850DAP, and J850FABRIX.