Learn More
Electron Microscopy Sciences Tough-Spots Labels on Rolls, 3/8", White
Supplier: Electron Microscopy Sciences 7758610
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: White