Learn More
Electron Microscopy Sciences Tough-Spots Labels on Rolls, 3/8", Assorted Colors
SDP
Catalog No. 501900052
Encompass_Preferred
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Assorted Colors
Catalog No. 50-190-0052 Supplier Electron Microscopy Sciences Supplier No. 7758611
Add to Cart
Add to Cart
Pre-cut, peel-off round labels for microcentrifuge tube tops on a roll. PVC labels accept writing from most lab markers including our Solvent Resistant Pens and Teeny Writers. A variety of sizes, colors and formats are available in a convenient dispensing box that assists in labeling.
Diameter: ⅜"
Color: Assorted Colors